Learn More
Biomatik Corporation Human Interferon Alpha/Beta Receptor 2 (Ifnar2) Elisa Kit
Supplier:  Biomatik Corporation EKC41647
Encompass_Preferred
Human Interferon Alpha/Beta Receptor 2 (Ifnar2) Elisa Kit for detecting Interferon Alpha/Beta Receptor 2(Ifnar2) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.8pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P48551; IFNAR2. Target Synonyms: Human interferon alpha/beta receptor; IFN alpha REC; IFN R; IFN-alpha binding protein; IFN-alpha/beta receptor 2; IFN-R-2; IFNABR; IFNAR2; IFNARB; IFNR; INAR2_HUMAN; Interferon (alpha beta and omega) receptor 2; Interferon alpha binding protein; Interferon alpha/beta receptor 2; Interferon alpha/beta receptor beta chain; Type I interferon receptor 2; Type I interferon receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4911