Learn More
Biomatik Corporation Dog Interleukin-13 (Il13) Elisa Kit
Supplier:  Biomatik Corporation EKC41671
Encompass_Preferred
Dog Interleukin-13 (Il13) Elisa Kit for detecting Interleukin 13 in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 12.5-800pg/ml. Sensitivity: 3.12pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9N0W9; IL13. Target Synonyms: IL13; Interleukin-13; IL-13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4915