Learn More
Biomatik Corporation Mouse Interleukin-20(IL20) ELISA kit
Supplier:  Biomatik Corporation EKC41691
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.3125ng/ml- 20ng/ml. Sensitivity: 0.078 ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q9JKV9. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-4920