Learn More
Biomatik Corporation Human Beta-Klotho (Klb) Elisa Kit
Supplier:  Biomatik Corporation EKC41719
Encompass_Preferred
Human Beta-Klotho (Klb) Elisa Kit for detecting Klotho Beta in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q86Z14; KLB. Target Synonyms: Beta-klotho; BetaKlotho; BKL; KLB; KLOTB_HUMAN; Klotho beta like protein; Klotho beta-like protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4926