Learn More
Biomatik Corporation Human Lymphocyte Activation Gene 3 Protein (Lag3) Elisa Kit
Supplier:  Biomatik Corporation EKC41730
Encompass_Preferred
Human Lymphocyte Activation Gene 3 Protein (Lag3) Elisa Kit for detecting Lymphocyte-Activation Gene 3 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P18627; LAG3. Target Synonyms: CD223; CD223 antigen; FDC protein; LAG-3; Lag3; LAG3_HUMAN; Lymphocyte activating 3; Lymphocyte activation gene 3; Lymphocyte activation gene 3 protein; Protein FDC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4930