Learn More
Biomatik Corporation Human Macrophage Scavenger Receptor Types I And Ii (Msr1) Elisa Kit
Supplier:  Biomatik Corporation EKC41789
Encompass_Preferred
Human Macrophage Scavenger Receptor Types I And Ii (Msr1) Elisa Kit for detecting Macrophage Scavenger Receptor 1 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5000pg/ml. Sensitivity: 19.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P21757; MSR1. Target Synonyms: MSR1; SCARA1; Macrophage scavenger receptor types I and II; Macrophage acetylated LDL receptor I and II; Scavenger receptor class A member 1; CD antigen CD204. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4938