Learn More
Biomatik Corporation Human Mimecan (Ogn) Elisa Kit
Supplier:  Biomatik Corporation EKC41819
Encompass_Preferred
Human Mimecan (Ogn) Elisa Kit for detecting Osteoglycin in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P20774; OGN. Target Synonyms: Corneal keratan sulfate proteoglycan; DKFZP586P2421; MIME_HUMAN; Mimecan; Mimecan proteoglycan; OG; OGN; OIF; Osteoglycin; Osteoinductive factor; SLRR3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4946