Learn More
Biomatik Corporation Human Peroxiredoxin 2 (Prdx2) Elisa Kit
Supplier:  Biomatik Corporation EKC41851
Encompass_Preferred
Human Peroxiredoxin 2 (Prdx2) Elisa Kit for detecting Peroxiredoxin 2 (Prdx2) in serum, plasma, tissue homogenates, cell lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 47-3000pg/ml. Sensitivity: 11.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P32119 ; PRDX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4953