Learn More
Biomatik Corporation Human Regenerating Islet-Derived Protein 3-Alpha (Reg3A) Elisa Kit
Supplier:  Biomatik Corporation EKC41870
Encompass_Preferred
Human Regenerating Islet-Derived Protein 3-Alpha (Reg3A) Elisa Kit for detecting Regenerating Islet-Derived Protein 3-Alpha(Reg3A) in serum, plasma, tissue homogenates, cell culture supernates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q06141; REG3A. Target Synonyms: FLJ18565; Hepatocarcinoma intestine pancreas; Hepatointestinal pancreatic protein; HIP; HIP/PAP; Human proislet peptide; INGAP; Pancreatic beta cell growth factor; Pancreatitis associated protein 1; Pancreatitis associated protein; Pancreatitis-associated protein 1; PAP; PAP H; PAP homologous protein; PAP1; PBCGF; Proliferation inducing protein 34; Proliferation inducing protein 42; Reg III alpha; REG III; Reg III-alpha; REG-3-alpha; REG3; Reg3a; REG3A_HUMAN; Regenerating islet derived 3 alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4955