Learn More
Biomatik Corporation Mouse Reelin(RELN) ELISA kit
Supplier:  Biomatik Corporation EKC41876
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 6.25 pg/mL-400 pg/mL. Sensitivity: 1.56 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Q60841. Target Synonyms: PRO1598, RL, OTTHUMP00000211830. Restriction: For Research Use Only.
Catalog No. 50-229-4957