Learn More
Biomatik Corporation Human R-Spondin-1 (Rspo1) Elisa Kit
Supplier:  Biomatik Corporation EKC41885
Encompass_Preferred
Human R-Spondin-1 (Rspo1) Elisa Kit for detecting R-Spondin Homolog (Xenopus Laevis) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4000pg/ml. Sensitivity: 15.6pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q2MKA7; RSPO1. Target Synonyms: CRISTIN3; FLJ40906; hRspo1; R spondin homolog; R spondin homolog (Xenopus laevis); R spondin1; R-spondin-1; Roof plate specific spondin; Roof plate-specific spondin-1; RP11-566C13.1; RSPO; Rspo1; RSPO1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4960