Learn More
Biomatik Corporation Human Thrombospondin-2 (Thbs2) Elisa Kit
Supplier:  Biomatik Corporation EKC41969
Encompass_Preferred
Human Thrombospondin-2 (Thbs2) Elisa Kit for detecting Thrombospondin 2 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 1.562-100ng/ml. Sensitivity: 1.191ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P35442; THBS2. Target Synonyms: THBS2; Thrombospondin-2; TSP2; TSP2_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4975