Learn More
Biomatik Corporation Human Twisted Gastrulation Protein Homolog 1 (Twsg1) Elisa Kit
Supplier:  Biomatik Corporation EKC42004
Encompass_Preferred
Human Twisted Gastrulation Protein Homolog 1 (Twsg1) Elisa Kit for detecting Twisted Gastrulation Homolog 1 (Drosophila) in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 125-8000pg/ml. Sensitivity: 31.25pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q9GZX9; TWSG1. Target Synonyms: PSEC0250; twisted gastrulation homolog 1 (Drosophila); TSG; TSG1; Twisted gastrulation BMP signaling modulator 1; Twisted gastrulation homolog 1; Twisted gastrulation protein; Twisted gastrulation protein homolog 1; TWSG1; TWSG1_HUMAN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4979