Learn More
Biomatik Corporation Mouse Stromal Cell Derived Factor 1Α (Sdf-1Α/Sdf1A) Elisa Kit
Supplier:  Biomatik Corporation EKC42063
Encompass_Preferred
Mouse Stromal Cell Derived Factor 1Α (Sdf-1Α/Sdf1A) Elisa Kit for detecting Stromal Cell Derived Factor 1Α(Sdf-1Α/Sdf1A) in serum, plasma, cell culture supernates, tissue homogenates. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P40224; Cxcl12. Target Synonyms: Cxcl12; Sdf1; Stromal cell-derived factor 1; SDF-1; 12-O-tetradecanoylphorbol 13-acetate repressed protein 1; TPAR1; C-X-C motif chemokine 12; Pre-B cell growth-stimulating factor; PBSF; Thymic lymphoma cell-stimulating factor; TLSF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4989