Learn More
Biomatik Corporation Horse Tri-Iodothyronine (T3) Elisa Kit
Supplier:  Biomatik Corporation EKC42070
Encompass_Preferred
Horse Tri-Iodothyronine (T3) Elisa Kit for detecting Tri-Iodothyronine (T3) in serum, plasma, tissue homogenates. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 0.5-8ng/ml. Sensitivity: 0.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-4991