Learn More
Biomatik Corporation Human Fibroblast Growth Factor 6 (Fgf6) Elisa Kit
Supplier:  Biomatik Corporation EKC42209
Encompass_Preferred
Human Fibroblast Growth Factor 6 (Fgf6) Elisa Kit for detecting Fibroblast Growth Factor 6 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 7.81-500pg/ml. Sensitivity: 1.95pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P10767; FGF6. Target Synonyms: FGF 6; FGF-6; Fgf6; FGF6_HUMAN; Fibroblast growth factor 6; Fibroblast growth factor 6 precursor; HBGF 6; HBGF-6; HBGF6; Heparin secretory-transforming protein 2; Heparin-binding growth factor 6; HST 2; HST-2; HST2; HSTF-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5004