Learn More
Biomatik Corporation Mouse Granulocyte Colony Stimulating Factor,G-CSF ELISA Kit
Supplier:  Biomatik Corporation EKC42211
Encompass_Preferred
Sample Type: serum, plasma,tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 7.8 pg/mL-500 pg/mL. Sensitivity: 1.95 pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P09920. Target Synonyms: G-CSF, GCSF, MGC45931, colony stimulating factor 3|filgrastim|granulocyte colony stimulating factor|lenograstim|pluripoietin. Restriction: For Research Use Only.
Catalog No. 50-229-5006