Learn More
Biomatik Corporation Mouse L-Selectin ELISA kit
Supplier:  Biomatik Corporation EKC42214
Encompass_Preferred
Sample Type: serum, plasma, tissue homogenates. Species Reactivity: Mus musculus (Mouse). Detection Range: 0.156 ng/mL-10 ng/mL. Sensitivity: 0.005 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: P18337. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5009