Learn More
Biomatik Corporation Rabbit Corticosterone,CORT ELISA Kit
Supplier:  Biomatik Corporation EKC42232
Encompass_Preferred
Sample Type: serum, plasma. Species Reactivity: Oryctolagus cuniculus (Rabbit). Detection Range: 0.5 ng/mL-20 ng/mL. Sensitivity: 0.5 ng/mL. Format: 96-Well Plate. Assay Type: Not Available. Detection Method: Colorimetric. Assay Time: 1-5 hours. UniProt ID: Not Available. Target Synonyms: Not Available. Restriction: For Research Use Only.
Catalog No. 50-229-5021