Learn More
Biomatik Corporation Human Macrophage Inflammatory Protein 5, Mip-5 Elisa Kit
Supplier:  Biomatik Corporation EKC42247
Encompass_Preferred
Human Macrophage Inflammatory Protein 5, Mip-5 Elisa Kit for detecting Chemokine (C-C Motif) Ligand 15 in serum, plasma, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 7.81pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: Q16663; CCL15. Target Synonyms: C-C motif chemokine 15; C-C motif chemokine ligand 15; CC chemokine 3; CCL15; CCL15 (29-92); CCL15_HUMAN; Chemokine (C C motif) ligand 15; Chemokine CC-2; Chemokine CC2; HCC-2; HCC2; HMRP 2B; HMRP2B; Leukotactin 1; Leukotactin-1; LKN-1; LKN1; Macrophage inflammatory protein 5; MIP 1D; MIP-1 delta; MIP-1D; MIP-5; MIP1 delta; MIP1d; MIP5; MRP 2B; Mrp-2b; Mrp2b; NCC 3; NCC-3; NCC3; New CC chemokine 3; OTTHUMP00000163955; SCYA15; SCYL3; Small inducible cytokine A15; Small inducible cytokine subfamily A (Cys Cys); member 15; Small-inducible cytokine A15; SY15. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-5025