Learn More
Biomatik Corporation Sheep Resistin (RETN) ELISA Kit
Supplier:  Biomatik Corporation EKU11398
Encompass_Preferred
Sheep Resistin (RETN) ELISA Kit for detecting RETN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Sheep (Ovis aries). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A0A068JAR6. Gene ID: 101122857. Target Synonyms: ADSF; FIZZ3; RETN1; RSTN; XCP1; Adipose tissue-specific secretory factor; C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein; Cysteine-rich secreted FIZZ3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3285