Learn More
Biomatik Corporation Human Surfactant Associated Protein A3 (SPA3) ELISA Kit
Supplier:  Biomatik Corporation EKU11448
Encompass_Preferred
Human Surfactant Associated Protein A3 (SPA3) ELISA Kit for detecting SPA3 in serum, plasma, tissue homogenates, cell lysates, lung lavage fluid, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P0C7M3. Target Synonyms: SFTPH; SP-H; Surfactant-associated protein H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3291