Learn More
Biomatik Corporation Human Transforming Growth Factor Beta 2 (TGFb2) ELISA Kit
Supplier:  Biomatik Corporation EKU11478
Encompass_Preferred
Human Transforming Growth Factor Beta 2 (TGFb2) ELISA Kit for detecting TGFb2 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P61812. Gene ID: 7042. Target Synonyms: TGF-B2; TGF-Beta2; G-TSF; LAP; Cetermin; Polyergin; Latency-associated peptide; BSC-1 cell growth inhibitor; Glioblastoma-derived T-cell suppressor factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3295