Learn More
Biomatik Corporation Rat Wingless Type MMTV Integration Site Family, Member 5A (WNT5A) ELISA Kit
Supplier:  Biomatik Corporation EKU11539
Encompass_Preferred
Rat Wingless Type MMTV Integration Site Family, Member 5A (WNT5A) ELISA Kit for detecting WNT5A in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QXQ7. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3306