Learn More
Biomatik Corporation Mouse Von Willebrand Factor Cleaving Protease (vWFCP) ELISA Kit
Supplier:  Biomatik Corporation EKU11688
Encompass_Preferred
Mouse Von Willebrand Factor Cleaving Protease (vWFCP) ELISA Kit for detecting vWFCP in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q769J6. Gene ID: 279028. Target Synonyms: ADAMTS13; TTP; vWF-cp; vWF-cleaving protease; A Disintegrin And Metalloproteinase With A Thrombospondin Type 1 Motif Member 13. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3332