Learn More
Biomatik Corporation Human Alanine Glyoxylate Aminotransferase 2 (AGXT2) ELISA Kit
Supplier:  Biomatik Corporation EKU11751
Encompass_Preferred
Human Alanine Glyoxylate Aminotransferase 2 (AGXT2) ELISA Kit for detecting AGXT2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BYV1. Gene ID: 64902. Target Synonyms: AGT2; Beta-Alanine-Pyruvate Aminotransferase; (R)-3-amino-2-methylpropionate--pyruvate transaminase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3339