Learn More
Biomatik Corporation Human Alcohol Dehydrogenase 4 (ADH4) ELISA Kit
Supplier:  Biomatik Corporation EKU11767
Encompass_Preferred
Human Alcohol Dehydrogenase 4 (ADH4) ELISA Kit for detecting ADH4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08319. Gene ID: 127. Target Synonyms: Alcohol dehydrogenase class II pi chain. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3341