Learn More
Biomatik Corporation Human Pleckstrin (PLEK) ELISA Kit
Supplier:  Biomatik Corporation EKU11794
Encompass_Preferred
Human Pleckstrin (PLEK) ELISA Kit for detecting PLEK in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08567. Gene ID: 5341. Target Synonyms: P47; Platelet 47 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3346