Learn More
Biomatik Corporation Human Cystathionine Beta Synthase (CbS) ELISA Kit
Supplier:  Biomatik Corporation EKU11806
Encompass_Preferred
Human Cystathionine Beta Synthase (CbS) ELISA Kit for detecting CbS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35520. Gene ID: 102724560. Target Synonyms: HIP4; Beta-thionase; Serine sulfhydrase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3348