Learn More
Biomatik Corporation Human Family With Sequence Similarity 3, Member C (FAM3C) ELISA Kit
Supplier:  Biomatik Corporation EKU11819
Encompass_Preferred
Human Family With Sequence Similarity 3, Member C (FAM3C) ELISA Kit for detecting FAM3C in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92520. Gene ID: 10447. Target Synonyms: ILEI; Predicted Osteoblast Protein; Interleukin Like EMT Inducer. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3351