Learn More
Biomatik Corporation Human Cytoskeleton Associated Protein 4 (CKAP4) ELISA Kit
Supplier:  Biomatik Corporation EKU11842
Encompass_Preferred
Human Cytoskeleton Associated Protein 4 (CKAP4) ELISA Kit for detecting CKAP4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07065. Gene ID: 10970. Target Synonyms: P63; CLIMP-63; ERGIC-63; 63-kDa cytoskeleton-linking membrane protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3355