Learn More
Biomatik Corporation Human Tumor Necrosis Factor Receptor Superfamily, Member 8 (TNFRSF8) ELISA Kit
Supplier:  Biomatik Corporation EKU11922
Encompass_Preferred
Human Tumor Necrosis Factor Receptor Superfamily, Member 8 (TNFRSF8) ELISA Kit for detecting TNFRSF8 in serum, plasma, urine, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P28908. Gene ID: 943. Target Synonyms: CD30; KI-1; CD30L receptor; Ki-1 antigen; Lymphocyte activation antigen CD30. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3372