Learn More
Biomatik Corporation Human/Mouse/Rat/Rabbit/Dog/Bovine Glucagon Like Peptide 1 (GLP1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11928
Encompass_Preferred
Human/Mouse/Rat/Rabbit/Dog/Bovine Glucagon Like Peptide 1 (GLP1) Mini sample ELISA Kit for detecting GLP1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus), Rabbit (Oryctolagus cuniculus), Dog (Canis familiaris; Canine), Bovine (Bos taurus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.64pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01275. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3375