Learn More
Biomatik Corporation Human Adenylate Cyclase 3 (ADCY3) ELISA Kit
Supplier:  Biomatik Corporation EKU11961
Encompass_Preferred
Human Adenylate Cyclase 3 (ADCY3) ELISA Kit for detecting ADCY3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60266. Gene ID: 109. Target Synonyms: AC3; ATP pyrophosphate-lyase 3; Adenylate cyclase, olfactive type; Adenylate cyclase type III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3387