Learn More
Biomatik Corporation Mouse Drebrin 1 (DBN1) ELISA Kit
Supplier:  Biomatik Corporation EKU11996
Encompass_Preferred
Mouse Drebrin 1 (DBN1) ELISA Kit for detecting DBN1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QXS6. Gene ID: 56320. Target Synonyms: Developmentally-regulated brain protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3396