Learn More
Biomatik Corporation Rabbit Procollagen III (PCIII) ELISA Kit
Supplier:  Biomatik Corporation EKU12014
Encompass_Preferred
Rabbit Procollagen III (PCIII) ELISA Kit for detecting PCIII in serum, plasma, other biological fluids. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Target Synonyms: PCOL-III; PC-III; PC3; Type III Procollagen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3400