Learn More
Biomatik Corporation Mouse Solute Carrier Family 7, Member 11 (SLC7A11) ELISA Kit
Supplier:  Biomatik Corporation EKU12037
Encompass_Preferred
Mouse Solute Carrier Family 7, Member 11 (SLC7A11) ELISA Kit for detecting SLC7A11 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WTR6. Gene ID: 26570. Target Synonyms: CCBR1; xCT; Cystine/Glutamate Transporter,Cationic Amino Acid Transporter,y+ System; Amino acid transport system xc-; Calcium channel blocker resistance protein CCBR1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3406