Learn More
Biomatik Corporation Mouse Interleukin 1 Receptor Like Protein 1 (IL1RL1) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12067
Encompass_Preferred
Mouse Interleukin 1 Receptor Like Protein 1 (IL1RL1) Mini sample ELISA Kit for detecting IL1RL1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14719. Gene ID: 17082. Target Synonyms: T1; IL33R; ST2; DER4; FIT-1; ST2L; ST2V; Homolog Of Mouse Growth Stimulation-Expressed. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3412