Learn More
Biomatik Corporation Rat Glucocorticoid Receptor Alpha (GRa) ELISA Kit
Supplier:  Biomatik Corporation EKU12151
Encompass_Preferred
Rat Glucocorticoid Receptor Alpha (GRa) ELISA Kit for detecting GRa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.118ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P06536. Gene ID: 24413. Target Synonyms: GR-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3435