Learn More
Biomatik Corporation Human Pyruvate Dehydrogenase Kinase 4 (PDK4) ELISA Kit
Supplier:  Biomatik Corporation EKU12163
Encompass_Preferred
Human Pyruvate Dehydrogenase Kinase 4 (PDK4) ELISA Kit for detecting PDK4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16654. Gene ID: 5166. Target Synonyms: [Pyruvate dehydrogenase (acetyl-transferring) ] kinase isozyme 4, mitochondrial; Pyruvate Dehydrogenase Kinase Isozyme 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3438