Learn More
Biomatik Corporation Pig Angiopoietin Like Protein 3 (ANGPTL3) ELISA Kit
Supplier:  Biomatik Corporation EKU12182
Encompass_Preferred
Pig Angiopoietin Like Protein 3 (ANGPTL3) ELISA Kit for detecting ANGPTL3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F1S7Z7. Target Synonyms: ANGPT5; ANG-5; Angiopoietin 5; Angiopoietin-related protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3446