Learn More
Biomatik Corporation Pig Aggrecan (AGC) ELISA Kit
Supplier:  Biomatik Corporation EKU12187
Encompass_Preferred
Pig Aggrecan (AGC) ELISA Kit for detecting AGC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.78-50ng/ml. Sensitivity: 0.38ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q29011. Target Synonyms: ACAN; AGC1; AGCAN; CSPG1; CSPCP; CSPGCP; MSK16; SEDK; Cartilage-specific proteoglycan core protein; Large Aggregating Proteoglycan; Chondroitin Sulfate Proteoglycan 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3448