Learn More
Biomatik Corporation Human Eukaryotic Translation Initiation Factor 2 Alpha Kinase 3 (EIF2aK3) ELISA Kit
Supplier:  Biomatik Corporation EKU12234
Encompass_Preferred
Human Eukaryotic Translation Initiation Factor 2 Alpha Kinase 3 (EIF2aK3) ELISA Kit for detecting EIF2aK3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NZJ5. Gene ID: 9451. Target Synonyms: WRS; PEK; PERK; HsPEK; PRKR-like endoplasmic reticulum kinase; Pancreatic eIF2-alpha kinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3459