Learn More
Biomatik Corporation Human Ubiquitin Like Modifier Activating Enzyme 2 (UBA2) ELISA Kit
Supplier:  Biomatik Corporation EKU12238
Encompass_Preferred
Human Ubiquitin Like Modifier Activating Enzyme 2 (UBA2) ELISA Kit for detecting UBA2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBT2. Gene ID: 10054. Target Synonyms: SAE2; ARX; SUMO1 Activating Enzyme Subunit 2; Anthracycline-associated resistance ARX; Ubiquitin-like 1-activating enzyme E1B; SUMO-activating enzyme subunit 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3461