Learn More
Biomatik Corporation Rat Hepatocyte Nuclear Factor 1 Beta (HNF1b) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12243
Encompass_Preferred
Rat Hepatocyte Nuclear Factor 1 Beta (HNF1b) Mini sample ELISA Kit for detecting HNF1b in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23899. Gene ID: 25640. Target Synonyms: FJHN; TCF2; HNF1beta; HNF2; LFB3; LF-B3; MODY5; VHNF1; Transcription Factor 2,Hepatic; Variant Hepatic Nuclear Factor; Homeoprotein LFB3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3464