Learn More
Biomatik Corporation 3-Methoxytyramine (3-MT) ELISA Kit
Supplier:  Biomatik Corporation EKU12258
Encompass_Preferred
3-Methoxytyramine (3-MT) ELISA Kit for detecting 3-MT in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 61.7-5,000pg/ml. Sensitivity: 21.9pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: 3-Methoxy-4-Hydroxyphenethylamine. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3466