Learn More
Biomatik Corporation Spermidine (SMD) CLIA Kit
Supplier:  Biomatik Corporation EKU12266
Encompass_Preferred
Spermidine (SMD) CLIA Kit for detecting SMD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 781.25-200,000pg/ml. Sensitivity: 284.75pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3470