Learn More
Biomatik Corporation Human V-Ha-Ras Harvey Rat Sarcoma Viral Oncogene Homolog (HRAS) ELISA Kit
Supplier:  Biomatik Corporation EKU12285
Encompass_Preferred
Human V-Ha-Ras Harvey Rat Sarcoma Viral Oncogene Homolog (HRAS) ELISA Kit for detecting HRAS in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01112. Gene ID: 3265. Target Synonyms: HRAS1; K-Ras; N-Ras; RASH1; C-Bas/Has; H-Ras; Transforming protein p21. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3480