Learn More
Biomatik Corporation Human Discoidin Domain Receptor Family, Member 1 (DDR1) ELISA Kit
Supplier:  Biomatik Corporation EKU12343
Encompass_Preferred
Human Discoidin Domain Receptor Family, Member 1 (DDR1) ELISA Kit for detecting DDR1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08345. Gene ID: 780. Target Synonyms: CD167a; CAK; DDR; EDDR1; MCK10; NEP; NTRK4; PTK3; PTK3A; RTK6; TRKE; Cell adhesion kinase; Epithelial discoidin domain receptor 1; Mammary carcinoma kinase 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3495