Learn More
Biomatik Corporation Human Angiotensinogen (AGT) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU12368
Encompass_Preferred
Human Angiotensinogen (AGT) Mini sample ELISA Kit for detecting AGT in serum, plasma, urine, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.17ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01019. Gene ID: 183. Target Synonyms: ANHU; SERPINA8; Angiotensin; Renin Substrate; Serpin Peptidase Inhibitor,Clade A,Member 8 (Alpha-1 Antiproteinase,Antitrypsin). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-3501